
N-Methylpropionamide ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:1187-58-2
:
Formula:C₄H₉NO
MDL Number:MFCD00009300
:87.12 g/mol
Boiling Pt:147°C
Flash Pt:105°C
EINECS:214-699-7
:GHS07
Specification Test Results
- Appearance:Colorless clear liquid
- Purity (GC):min. 99.0 %
Specification Test Results
Frequently Bought Together