Order Entry
Spain
ContactUsLinkComponent
 
cis-2-Methylcyclohexanol ≥98.0% (by GC)
Catalog # TCIAM0351-5ML
Supplier:  TCI
CAS Number:  
undefined
cis-2-Methylcyclohexanol ≥98.0% (by GC)
Catalog # TCIAM0351-5ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 mL
  • Seq:
    2210

Specifications

Specification Test Results

  • Appearance:
    Colorless clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
 :114.19 g/mol
Boiling Pt:165°C
Melting Pt:25…27 °C
Density:0,936 g/cm³ (20 °C)
Flash Pt:58°C
EINECS:231-187-9
UN:2617
ADR:3,III
 :GHS02
 :GHS07
CAS Number:7443-70-1
Formula:C₇H₁₄O
MDL Number:MFCD00064169