D-α-Aminoadipic acid ≥95.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:7620-28-2
:
Formula:C₆H₁₁NO₄
MDL Number:MFCD00063118
:161.16 g/mol
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 95.0 %
- Purity (Nonaqueous Titration):min. 95.0 %
- Specific rotation [a]20/D:-20.0 to -27.0 deg (C=1, 6mol/L HCl)
Specification Test Results
Related Information
Frequently Bought Together