
Hexakis(bromomethyl)benzene ≥98.0% (by HPLC, titration analysis)
: TCI
:3095-73-6
:
:C₁₂H₁₂Br₆
:MFCD00182539
:635.65 g/mol
:221-443-8
:3261
:8,III
:GHS05
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Argentmetric Titration):min. 98.0 %
Frequently Bought Together