
4'-Hydroxy-2'-methylacetophenone ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:875-59-2
:
Formula:C₉H₁₀O₂
MDL Number:MFCD00002303
:150.18 g/mol
Boiling Pt:313°C
EINECS:212-874-2
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:128.0 to 132.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together