2,4-Pentanedione dioxime ≥98.0% (by total nitrogen basis)
Catalog # TCIAP0792-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:145.0 to 149.0 deg-C
- Solubility in hot Water:almost transparency
Specification Test Results
About this item
:
:130.15 g/mol
EINECS:218-472-3
CAS Number:2157-56-4
Formula:C₅H₁₀N₂O₂
MDL Number:MFCD00013932