2,6-Dichlorophenethylamine ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:14573-23-0
:
Formula:C₈H₉Cl₂N
MDL Number:MFCD00060612
:190.07 g/mol
Boiling Pt:129°C
:GHS07
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together