Order Entry
Spain
ContactUsLinkComponent
 
 
1-Chloro-2-phenylethane ≥99.0% (by GC)
  TCIAC0167-25G
 :  TCI
 :  
undefined
 
1-Chloro-2-phenylethane ≥99.0% (by GC)
  TCIAC0167-25G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 99.0 %

 

 

 :
 :140.61 g/mol
 :200°C
 :–60 °C
 :1,07 g/cm³ (20 °C)
 :64°C
 :Ambient
 :210-725-6
 :GHS07
 :622-24-2
 :ClCH₂CH₂C₆H₅
 :MFCD00000977