1-Phenyl-2-butanone ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:1007-32-5
:
Formula:C₆H₅C(O)CH₂CH₂CH₃
MDL Number:MFCD00009315
:148.2 g/mol
Boiling Pt:226°C
Melting Pt:5 °C
Density:0,989 g/cm³ (20 °C)
Flash Pt:90°C
EINECS:213-752-1
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 95.0 %
Specification Test Results
Frequently Bought Together