
3,4-Difluorobenzoic acid ≥98.0% (by GC, titration analysis)
: TCI
:455-86-7
:
:C₇H₄F₂O₂
:MFCD00011672
:158.1 g/mol
:242 °C (1013 hPa)
:119…122 °C
:1 g/cm³ (20 °C)
:Ambient
:207-249-6
:GHS07
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:121.0 to 124.0 deg-C
- Solubility in Methanol:very faint turbidity
Frequently Bought Together