2,4-Dichlorothiophenol ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:1122-41-4
:
Formula:C₆H₄Cl₂S
MDL Number:MFCD00041421
:179.07 g/mol
Boiling Pt:115°C
Melting Pt:22 °C
Density:1,44 g/cm³ (20 °C)
EINECS:214-346-7
:GHS07
Specification Test Results
- Appearance:White or Colorless to Light yellow powder to lump to clear liquid
- Purity (GC):min. 97.0 %
Specification Test Results
Frequently Bought Together