
2,6-Dichlorothiophenol ≥95.0% (by GC)
: TCI
:24966-39-0
:
:C₆H₄Cl₂S
:MFCD00004833
:179.07 g/mol
:246 °C (1013 hPa)
:45…50 °C
:Ambient
:416-720-7
:GHS07
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 95.0 %
- Melting point:47.0 to 51.0 deg-C
Frequently Bought Together