Order Entry
Spain
ContactUsLinkComponent
 
Irritant
3,4-Dichlorothiophenol ≥97.0% (by GC, titration analysis)
Catalog # TCIAD1697-5G
Supplier:  TCI
CAS Number:  
undefined
Irritant
3,4-Dichlorothiophenol ≥97.0% (by GC, titration analysis)
Catalog # TCIAD1697-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White or Colorless to Light orange to Yellow powder to lump to clear liquid
  • Purity (GC):
    min. 97.0 %
  • Purity (Iodometric Titration):
    min. 97.0 %
  • Freezing point:
    19.0 to 22.0 deg-C

Specification Test Results

About this item

 :
 :179.07 g/mol
Boiling Pt:81°C
Melting Pt:87…89 °C
Density:1,4175 g/cm³ (20 °C)
EINECS:227-485-3
 :GHS07
CAS Number:5858-17-3
Formula:C₆H₄Cl₂S
MDL Number:MFCD00004840