


2,4-Dichloroaniline ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:554-00-7
:
Formula:C₆H₅Cl₂N
MDL Number:MFCD00007661
:162.02 g/mol
Boiling Pt:242°C
Melting Pt:59…62 °C
Density:1,567 g/cm³ (20 °C)
Flash Pt:149 °C
Storage Temperature:Ambient
EINECS:209-057-8
UN:3442
ADR:6.1,II
:GHS06
:GHS08
:GHS09
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 99.0 %
- Melting point:62.0 to 65.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together