
3,4-Dichlorobenzonitrile ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:6574-99-8
:
Formula:C₇H₃Cl₂N
MDL Number:MFCD00016379
:172.01 g/mol
Melting Pt:86…89 °C
Storage Temperature:Ambient
EINECS:229-494-8
UN:3439
ADR:6.1,III
:GHS06
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:71.0 to 75.0 deg-C
Specification Test Results
Frequently Bought Together