
2,4-Dichlorobenzonitrile ≥98.0% (by GC)
Catalog # TCIAD3349-5G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:59.0 to 63.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
About this item
:
:172.01 g/mol
Boiling Pt:125°C
Melting Pt:52…53 °C
Storage Temperature:Ambient
EINECS:229-493-2
UN:3439
ADR:6.1,III
:GHS06
CAS Number:6574-98-7
Formula:C₇H₃Cl₂N
MDL Number:MFCD00016373