
Pyromellitic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:89-05-4
:
Formula:C₁₀H₆O₈
MDL Number:MFCD00002471
:254.15 g/mol
Merck Index:14,08004
Boiling Pt:516 °C (1013 hPa)
Melting Pt:272 °C
Density:1,79 g/cm³ (20 °C)
EINECS:201-879-5
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):min. 98.0 %
- Solubility in Methanol:very faint turbidity
Specification Test Results
Frequently Bought Together