Order Entry
Spain
ContactUsLinkComponent
 
Isopropyl benzoate ≥99.0% (by GC)
  TCIAB0073-25ML
 :  TCI
 :  
undefined
Isopropyl benzoate ≥99.0% (by GC)
  TCIAB0073-25ML
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 mL
  • Seq:
    2315

 

 

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 99.0 %

 

 

 :
 :164.2 g/mol
 :218°C
 :-26°C
 :–26°C
 :99°C
 :213-361-6
 :939-48-0
 :C₁₀H₁₂O₂
 :MFCD00048289