1,4-Dibromo-2-nitrobenzene ≥98.0% (by GC)
: TCI
:3460-18-2
:
:C₆H₃Br₂NO₂
:MFCD00007046
:280.9 g/mol
:81…85 °C
:2,374 g/cm³ (20 °C)
:222-404-8
:GHS07
:GHS08
- Appearance:White to Amber powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:82.0 to 86.0 deg-C
- Solubility in Methanol:very faint turbidity
Frequently Bought Together