
3-(Trifluoromethyl)phenylhydrazine ≥97.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:368-78-5
:
Formula:C₇H₇F₃N₂
MDL Number:MFCD00025093
:176.14
Boiling Pt:83°C
EINECS:206-713-5
:GHS07
Specification Test Results
- Appearance:Light orange to Yellow to Green clear liquid
- Purity (GC):min. 97.0 %
- Purity (Potassium iodate Method):min. 97.0 %
Specification Test Results
Frequently Bought Together