
1-Bromo-3,5-dichlorobenzene ≥98.0% (by GC)
Catalog # TCIAB2223-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:75.0 to 79.0 deg-C
Specification Test Results
About this item
:
:225.9 g/mol
Boiling Pt:227 °C (1013 hPa)
Melting Pt:82…84 °C
EINECS:243-270-7
:GHS07
CAS Number:19752-55-7
Formula:C₆H₃BrCl₂
MDL Number:MFCD00000584