
4-Bromobenzoic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:586-76-5
:
Formula:C₇H₅BrO₂
MDL Number:MFCD00002529
:201.02 g/mol
Merck Index:14,01408
Boiling Pt:300 °C (1013 hPa)
Melting Pt:252…256 °C
Density:1,894 g/cm³ (20 °C)
EINECS:209-581-7
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Solubility in hot Methanol:almost transparency
Specification Test Results
Related Information
Frequently Bought Together