Order Entry
Spain
ContactUsLinkComponent
 
3-Methyl-1-hexene ≥99.0% (by GC)
  TCIAM0301-5ML
 :  TCI
 :  
undefined
3-Methyl-1-hexene ≥99.0% (by GC)
  TCIAM0301-5ML
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    5 mL
  • Seq:
    2210
  • Environmentally Preferable:

 

 

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 99.0 %

 

 

 :
 :98.19 g/mol
 :222-283-1
 :3295
 :GHS02
 :GHS08
 :Ambient
 :3404-61-3
 :C₇H₁₄
 :MFCD00009407
 :84°C
 :-122
 :–18°C
 :3 II