Order Entry
Spain
ContactUsLinkComponent
1-Eicosene ≥97.0% (by GC)
Health
1-Eicosene ≥97.0% (by GC)
Catalog # TCIAE0774-25ML
Supplier:  TCI
CAS Number:  
undefined
Health
1-Eicosene ≥97.0% (by GC)
Catalog # TCIAE0774-25ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 mL
  • Seq:
    2315

Specifications

Specification Test Results

  • Appearance:
    White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
  • Purity (GC):
    min. 97.0 %

Specification Test Results

About this item

 :
 :280.54 g/mol
Boiling Pt:151°C
Melting Pt:60…62 °C
Density:0,795 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:222-374-6
 :GHS08
CAS Number:3452-07-1
Formula:C₂₀H₄₀
MDL Number:MFCD00009013