Order Entry
Spain
ContactUsLinkComponent
 
Flammable
Health
2,2,5-Trimethylhexane ≥99.0% (by GC)
Catalog # TCIAT0475-5ML
Supplier:  TCI
CAS Number:  
undefined
Flammable
Health
2,2,5-Trimethylhexane ≥99.0% (by GC)
Catalog # TCIAT0475-5ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 mL
  • Seq:
    2210

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 99.0 %

Specification Test Results

About this item

 :
 :128.26 g/mol
Boiling Pt:122°C
Melting Pt:–105.8 °C
Density:0,7082 g/cm³ (20 °C)
Flash Pt:13°C
Storage Temperature:Ambient
EINECS:222-537-1
UN:1920
ADR:3,III
 :GHS02
 :GHS08
CAS Number:3522-94-9
Formula:C₉H₂₀
MDL Number:MFCD00026525