Order Entry
Spain
ContactUsLinkComponent
 
 
 
1-Chloro-2-methyl-2-propanol ≥98.0% (by GC)
  TCIAC0971-25ML
 :  TCI
 :  
undefined
 
 
1-Chloro-2-methyl-2-propanol ≥98.0% (by GC)
  TCIAC0971-25ML
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 mL
  • Seq:
    2315

 

 

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 98.0 %

 

 

 :
 :108.57 g/mol
 :124 °C (1013 hPa)
 :–20 °C
 :1,082 g/cm³ (0 °C)
 :38°C
 :Ambient
 :209-196-4
 :1987
 :3,III
 :GHS02
 :GHS07
 :558-42-9
 :C₄H₉ClO
 :MFCD00021807