Order Entry
Spain
ContactUsLinkComponent
 
3-Hydroxy-3-methylbutanone ≥95.0% (by GC)
 :  TCI
 
undefined
3-Hydroxy-3-methylbutanone ≥95.0% (by GC)
 :  TCI

 

  • Environmentally Preferable:

 

 

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 95.0 %

 

Frequently Bought Together