
1-Bromo-3,3-dimethylbutanone ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:5469-26-1
:
Formula:C₆H₁₁BrO
MDL Number:MFCD00000206
:179.06 g/mol
Boiling Pt:188°C
Melting Pt:76 °C
Density:1,3508 g/cm³ (0 °C)
Flash Pt:66°C
Storage Temperature:Refrigerator
EINECS:226-794-0
ADR:6.1,II
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 95.0 %
Specification Test Results
Frequently Bought Together