


1-Dodecanethiol ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:112-55-0
:
Formula:C₁₂H₂₆S
MDL Number:MFCD00004885
:202.4 g/mol
Boiling Pt:277°C
Melting Pt:–7°C
Density:0,855 g/cm³ (20 °C)
Flash Pt:128°C
Storage Temperature:Ambient
EINECS:203-984-1
UN:1760
ADR:8,III
:GHS05
:GHS07
:GHS09
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 95.0 %
Specification Test Results
Related Information
Frequently Bought Together