(±)-2-Nonanol ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:628-99-9
:
Formula:C₉H₂₀O
MDL Number:MFCD00004593
:144.26 g/mol
Boiling Pt:198°C
Melting Pt:–35 °C
Density:0,831 g/cm³ (20 °C)
Flash Pt:82°C
Storage Temperature:Ambient
EINECS:211-065-1
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together