Order Entry
Spain
ContactUsLinkComponent
Chloroform ≥99.5% (by GC) stabilised for HPLC
Toxic
Health
Environment
Chloroform ≥99.5% (by GC) stabilised for HPLC
Catalog # TCIAC1111-500ML
Supplier:  TCI
CAS Number:  
undefined
Toxic
Health
Environment
Chloroform ≥99.5% (by GC) stabilised for HPLC
Catalog # TCIAC1111-500ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

Specification Test Results

  • Appearance:
    Colorless clear liquid
  • Purity (GC):
    min. 99.5 %
  • Absorbance (250nm):
    max. 1.000
  • Absorbance (260nm):
    max. 0.150
  • Absorbance (270nm):
    max. 0.050

Specification Test Results

About this item

 :
 :119.38 g/mol
Merck Index:14,02141
Boiling Pt:61,7 °C (1013 hPa)
Melting Pt:-64°C
Melting Pt:–64°C
Density:1,4832 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-663-8
UN:1888
ADR:6.1,III
 :Stabilised with 2-methyl-2-butene (iso-amylene)
REACH:05-2114614906-45
 :GHS06
 :GHS08
 :GHS09
CAS Number:67-66-3
Formula:CHCl₃
MDL Number:MFCD00000826