


1,2-Dichloropropane ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:78-87-5
:
Formula:ClH₂CCH₂CH₂Cl
MDL Number:MFCD00000868
:112.99 g/mol
Merck Index:14,07854
Boiling Pt:97°C
Melting Pt:–100 °C
Density:1,158 g/cm³ (20 °C)
Flash Pt:14°C
Storage Temperature:Ambient
EINECS:201-152-2
UN:1279
ADR:3,II
:GHS02
:GHS07
:GHS08
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together