Sodium propionate ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:137-40-6
:
Formula:C₃H₅NaO₂
MDL Number:MFCD00002759
:96.06 g/mol
Merck Index:14,08669
Melting Pt:288.76 °C
Flash Pt:157 °C
EINECS:205-290-4
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Nonaqueous Titration):min. 98.0 %
- Solubility in Water:almost transparency
Specification Test Results
Frequently Bought Together