Order Entry
Spain
ContactUsLinkComponent
4-Octanone ≥98.0% (by GC)
Flammable
4-Octanone ≥98.0% (by GC)
Catalog # TCIAO0218-1ML
Supplier:  TCI
CAS Number:  
undefined
Flammable
4-Octanone ≥98.0% (by GC)
Catalog # TCIAO0218-1ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • Seq:
    2070

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
 :128.21 g/mol
Boiling Pt:166°C
Melting Pt:16.4…16.7 °C
Density:0,819 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:209-655-9
UN:1224
ADR:3,III
 :GHS02
CAS Number:589-63-9
Formula:C₈H₁₆O
MDL Number:MFCD00027241