3-Octanone ≥98.0% (by GC)
Supplier: TCI
About this item
- CAS Number:106-68-3
- Formula:C₈H₁₆O
- MDL Number:MFCD00009515
- Molecular Weight:128.21 g/mol
- Merck Index:14,06753
- Boiling Pt:167°C
- Melting Pt:–18.5 °C
- Density:0,82 g/cm³ (20 °C)
- Flash Pt:54°C
- Storage Temperature:Refrigerator
- EINECS:203-423-0
- UN:2271
- ADR:3,III
2 Options Available Below
Product Details & Documents
- CAS Number:106-68-3
- Formula:C₈H₁₆O
- MDL Number:MFCD00009515
- Molecular Weight:128.21 g/mol
- Merck Index:14,06753
- Boiling Pt:167°C
- Melting Pt:–18.5 °C
- Density:0,82 g/cm³ (20 °C)
- Flash Pt:54°C
- Storage Temperature:Refrigerator
- EINECS:203-423-0
- UN:2271
- ADR:3,III
Specifications for Product Family
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



