Order Entry
Spain
ContactUsLinkComponent
 
 
2-Tridecanone ≥95.0% (by GC)
  TCIAT0415-25G
 :  TCI
 :  
undefined
 
2-Tridecanone ≥95.0% (by GC)
  TCIAT0415-25G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Appearance:
    White or Colorless to Light orange to Yellow powder to lump to clear liquid
  • Purity (GC):
    min. 95.0 %

 

 

 :
 :198.35 g/mol
 :263°C
 :25…27 °C
 :0,82 g/cm³ (20 °C)
 :Ambient
 :209-784-0
 :3077
 :9,III
 :GHS09
 :593-08-8
 :C₁₃H₂₆O
 :MFCD00008968