Order Entry
Spain
ContactUsLinkComponent
2,4-Dinitro-1-naphthol ≥98.0% (by HPLC, titration analysis)
Irritant
2,4-Dinitro-1-naphthol ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAD0839-25G
Supplier:  TCI
CAS Number:  
undefined
Irritant
2,4-Dinitro-1-naphthol ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAD0839-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Yellow to Orange powder to crystaline
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Neutralization titration):
    min. 98.0 %
  • Melting point:
    133.0 to 139.0 deg-C
  • Lambda max.:
    367.0 to 373.0 nm (MeOH)

Specification Test Results

About this item

 :
 :234.17 g/mol
Boiling Pt:408 °C (1013 hPa)
Melting Pt:130…133 °C
Storage Temperature:Ambient
EINECS:210-093-1
UN:3077
ADR:9,III
 :GHS07
CAS Number:605-69-6
Formula:C₁₀H₆N₂O₅
MDL Number:MFCD00065352