

Valeraldehyde ≥95.0% (by GC)
: TCI
:110-62-3
:
:C₅H₁₀O
:MFCD00007026
:14,09902
:102°C
:–92 °C
:0,81 g/cm³ (20 °C)
:12°C
:2058
:3,II
:86.13 g/mol
:203-784-4
:GHS02
:GHS07
:Refrigerator
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 95.0 %
Frequently Bought Together