

1-Bromo-4-chlorobutane ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:6940-78-9
:
Formula:BrH₂C(CH₂)₂CH₂Cl
MDL Number:MFCD00001010
:171.46 g/mol
Boiling Pt:81°C
Density:1,4895 g/cm³ (20 °C)
Flash Pt:60°C
Storage Temperature:Ambient
EINECS:230-089-3
UN:1993
ADR:3,III
:GHS02
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 99.0 %
Specification Test Results
Frequently Bought Together