(±)-3-Methylcyclohexanone ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:591-24-2
:
Formula:C₇H₁₂O
MDL Number:MFCD00001639
Boiling Pt:169°C
Molecular Weight:112.17 g/mol
Melting Pt:-73
Flash Pt:48°C
Storage Temperature:Ambient
EINECS:209-710-7
UN:2297
ADR:3 III
:GHS02
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 97.0 %
Specification Test Results
Frequently Bought Together