Order Entry
Spain
Orders LinkContactUsLinkComponent
Thymolphthalein

Thymolphthalein

Supplier:   TCI
Thymolphthalein
Supplier:  TCI
Certificates
Thymolphthalein

2 Options Available Below

Product Details & Documents
Synonyms:  3,3-Bis(4-hydroxy-5-isopropyl-o-tolyl)phthalide, 3,3-Bis(4-hydroxy-5-isopropyl-2-methylphenyl)isobenzofuran-1(3H)-one
  • CAS Number:125-20-2
  • Formula:C₂₈H₃₀O₄
  • MDL Number:MFCD00005909
  • Molecular Weight:430.54 g/mol
  • Merck Index:14,09401
  • Melting Pt:253 °C
  • Storage Temperature:Ambient
  • EINECS:204-729-7
Specifications for Product Family

Specification Test Results

  • Appearance:
    White to Orange to Green powder to crystal
  • Absorbance:
    min. 0.350 (5mg/L, pH10.5, 592.0 to 596.0 nm)
  • Solubility in EtOH (50vol%):
    transparency
  • Drying loss:
    max. 1.0 %
  • Transition interval (pH):
    (colorless)8.6 to 10.5 (blue)
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 17,00 €
    17,00 €
    1 * 1 g
       17,00 €

  • Glass bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 46,00 €
    46,00 €
    1 * 25 g
       46,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.