
trans-2-Pentenoic acid ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:13991-37-2
:
Formula:C₅H₈O₂
MDL Number:MFCD00002704
:100.12 g/mol
Boiling Pt:97°C
Density:0,9903 g/cm³ (20 °C)
Flash Pt:102°C
EINECS:237-791-9
UN:3265
ADR:8,III
:GHS05
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 95.0 %
Specification Test Results
Frequently Bought Together