L(+)-Ribose ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:24259-59-4
:
Formula:C₅H₁₀O₅
MDL Number:MFCD00167010
:150.13 g/mol
Boiling Pt:331 °C (1013 hPa)
Melting Pt:86…87 °C
Storage Temperature:Ambient
EINECS:246-110-4
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Melting point:88.0 to 92.0 deg-C
- Specific rotation [a]20/D:+19.0 to +21.0 deg (C=1, H2O)
Specification Test Results
Frequently Bought Together