Order Entry
Spain
ContactUsLinkComponent
 
Safranine O
 :  TCI
 
Safranine O
 :  TCI

 

  • Appearance:
    Orange to Brown to Dark red powder to crystal
  • Lambda max.:
    517.0 to 522.0 nm (H2O)
  • Absorbance (E1%1cm):
    min. 550 (H2O, 517.0 to 522.0 nm) (calcd.on dried substance)
  • Drying loss:
    max. 15.0 %

 

Frequently Bought Together