Order Entry
Spain
ContactUsLinkComponent
 
 
2-Methyl-4-pentenoic acid ≥98.0% (by GC, titration analysis)
  TCIAM1143-10ML
 :  TCI
 :  
undefined
 
2-Methyl-4-pentenoic acid ≥98.0% (by GC, titration analysis)
  TCIAM1143-10ML
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • Seq:
    2275

 

 

  • Appearance:
    Colorless to Red to Green clear liquid
  • Purity (GC):
    min. 98.0 %
  • Purity (Neutralization titration):
    min. 98.0 %

 

 

 :
 :114.14 g/mol
 :104°C
 :0,95 g/cm³ (20 °C)
 :107°C
 :216-404-7
 :3265
 :8,III
 :GHS05
 :1575-74-2
 :C₆H₁₀O₂
 :MFCD00015570