2-Deoxy-D(-)-ribose ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
Synonyms: 2-Deoxy-D(-)-erythropentose, Thyminose, 2-Deoxy-D(-)-arabinose, 2'-Deoxy-D-Ribose, (3S,4R)-3,4,5-Trihydroxypentanal
CAS Number:533-67-5
:
Formula:C₅H₁₀O₄
MDL Number:MFCD00135904
:134.13 g/mol
Merck Index:14,02908
Boiling Pt:292 °C (1013 hPa)
Melting Pt:95…97 °C
Storage Temperature:Ambient
EINECS:208-573-0
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC) (CAD):min. 98.0 area%
- Specific rotation [a]20/D:-54.0 to -58.0 deg (C=1, H2O)
- Solubility in Water:almost transparency
Specification Test Results
Frequently Bought Together