
2-(4-Iodophenyl)-3-(4-nitrophenyl)-5-phenyltetrazolium chloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:146-68-9
:
Formula:C₁₉H₁₃ClIN₅O₂
MDL Number:MFCD00011961
:505.7 g/mol
Melting Pt:227…229 °C
EINECS:205-676-2
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Yellow to Orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Argentmetric Titration):min. 98.0 % (after drying)
- Drying loss:max. 6.0 %
Specification Test Results
Frequently Bought Together