Undec-10-enoic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:112-38-9
:
Formula:C₁₁H₂₀O₂
MDL Number:MFCD00004442
:184.28 g/mol
Merck Index:14,09848
Boiling Pt:275°C
Melting Pt:24 °C
Density:0,91 g/cm³ (20 °C)
Flash Pt:146 °C
EINECS:203-965-8
:GHS07
Specification Test Results
- Appearance:White or Colorless to Light orange to Yellow powder to lump to clear liquid
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
Frequently Bought Together