


N,N-Diethylmethylamine ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:616-39-7
:
Formula:C₅H₁₃N
MDL Number:MFCD00009049
:87.16 g/mol
Boiling Pt:63 °C (1013 hPa)
Melting Pt:–196 °C
Density:0,7227 g/cm³ (20 °C)
Flash Pt:16°C
EINECS:210-480-5
UN:2733
ADR:3,II
:GHS02
:GHS05
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 97.0 %
Specification Test Results
Frequently Bought Together