L(+)-Lysine monohydrochloride ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:657-27-2
:
Formula:H₂N(CH₂)₄CH(NH₂)CO₂H·HCl
MDL Number:MFCD00064564
Merck Index:14,05636
Melting Pt:263…264 °C
:182.65 g/mol
Storage Temperature:Ambient
EINECS:211-519-9
Specification Test Results
- Appearance:White powder to crystal
- Purity (Nonaqueous Titration):min. 98.0 %
- Specific rotation [a]20/D:+19.0 to +21.5 deg (C=8, HCl (1+1))
- Solubility in Water:almost transparency
- Related compounds (TLC):to pass test
Specification Test Results
Related Information
Frequently Bought Together